Would you like to log in again?
Apoptosis and Necrosis Quantitation Kits, Biotium
Supplier: Biotium
30018 30060 30017 30061
89138-930EA 299.83 USD InStock
89138-930 89138-978 89138-932 89138-976
Apoptosis and Necrosis Quantitation Kits, Biotium
Assays Cellular Assays
Biotium's apoptosis/necrosis quantitation kits pair green fluorescent FITC-Annexin V conjugate with the membrane impermeant red fluorescent nucleic acid stain ethidium homodimer III
Green fluorescent CF™488A is brighter and more photostable than FITC, and unlike FITC, CF™488 fluorescence is not pH sensitive. Biotium offers CF™488A Annexin V conjugate in combination with propidium iodide or 7-AAD, membrane impermeant nucleic acid vital dyes that stain dead cell with red fluorescence. Propidium iodide is recommended for flow cytometry, while 7-AAD is recommended for fluorescence microscopy due to minimal fluorescence spill-over of 7-AAD in the green channel.
Features:
Fluorescently stain apoptotic cells green and necrotic cells red.
CF™488A Annexin V and other Annexin V conjugates of Biotium's bright and photostable CF™ Dyes also are available as stand-alone products. See catalog number 89138-858.
In the Apoptosis and Necrosis Quantitation Kit, FITC-Annexin V stains phosphatidylserine on the surface of apoptotic cells, while ethidium homodimer III stains necrotic and late apoptotic cells that have compromised plasma membrane integrity. Ethidium homodimer III also allows visualization of dead cell nuclear morphology to distinguish late apoptotic cells from necrotic cells. The Apoptotic, Necrotic and Healthy cells Quantitation Kit includes FITC-Annexin V, ethidium homodimer III, and Hoechst 33342 DNA dye, which stain the nuclei of all cells with blue fluorescence. The kits can be used for detection by microplate reader, flow cytometry, or fluorescence microscopy.
Ordering information: Kit size based on flow cytometry assay. See product datasheets for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty