Would you like to log in again?
Adapter, Thermocouple with 1:5 PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
5272-15 5272-17 5274-43 5274-22
10098-748EA 204.3 USD InStock
10098-748 89051-548 89051-546 89051-544
Adapter, Thermocouple with 1:5 PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Thermocouple adapter with 1:5 PTFE stopcock with hose connection.
  • Stopcock with hose connection
  • Fabricated from borosilicate glass
Supplied with bushing and FETFE O-ring which allows for compression seal with probes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty