Would you like to log in again?
Adapter, Cap, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
8273-05
89083-174EA 36.37 USD InStock
89083-174
Adapter, Cap, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features an o-ring joint on one side and a blank cap on the other.
  • O-ring joint
  • Blank cap
  • Fabricated from borosilicate glass
Adapter has a 15mm o-ring joint (No. 15) on one side and a blank cap on the other.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty