Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adapter, Thermometer, [ST]10/20, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

8299-10
89062-926EA 37.75 USD InStock
89062-926
Adapter, Thermometer, [ST]10/20, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This thermometer adapter is made of PTFE with a knurled nut to adapt standard chemical thermometers.

  • With knurled nut
  • Made of PTFE
  • Maximum temperature of 160°C

Knurled nut adapts standard chemical thermometers with an O.D. up to 6mm. For use in standard taper 10/18 or 10/30 outer joints. Simply insert the thermometer to the desired depth and tighten the nut.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty