Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Farnesol (mixture of isomers)

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Farnesol2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-3,7,11-Trimethyldodeca-2,6,10-trien-1-olTRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL

F3035-5ML F3035-25ML
700002-832EA 64.37 USD InStock
700002-832 700002-830
Farnesol (mixture of isomers)
Farnesol (mixture of isomers)
Farnesol, Mixture of Isomers, a 15-carbon organic compound, is used in perfumery and present in many essential oils such as neroli, rose, citronella, and musk.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Density: 0.888
Storage Temperature: Ambient
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty