Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Lithium oxalate

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Lithium oxalateDilithium oxalateLithiumoxalate

L1122-25GM L1122-100GM
700005-750EA 597 USD InStock
700005-750 700005-752
di-Lithium oxalate
di-Lithium oxalate
Lithium Oxalate is a personal care products ingredient for items including cosmetics, shampoos, perfumes, soaps, lotions, and toothpastes.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₂Li₂O₄
MW: 101.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040596
CAS Number: 553-91-3
EINECS: 209-054-1
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty