Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Threonine ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

T3805-100GM T3805-25GM T3805-1KG
700007-504EA 2181.78 USD InStock
700007-504 700007-506 700007-502
DL-Threonine ≥98%
DL-Threonine
DL-Threonine, also known as 2-Amino-3-hydroxybutanoic acid, is an essential amino acid that is classified as polar and susceptible to several post-translational modifications. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063722
CAS Number: 80-68-2
EINECS: 201-300-6
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty