Would you like to log in again?
Purple Starchy:Purple Sweet:Yellow Starchy:Yelllow Sweet 1:1:1:1 Ratio
Supplier: CAROLINA BIOL SUPPLY CO SE
WF-17-6602
470017-836EA 22.65 USD InStock
470017-836
Purple Starchy:Purple Sweet:Yellow Starchy:Yelllow Sweet 1:1:1:1 Ratio
Specimens Live Specimens Plants - Living Material
Purple Starchy: Purple Sweet: Yellow Starchy: Yelllow Sweet. The result of test crossing the F1 hybrid with the recessive parent (R/r Su/su and r/r su/su).
  • Genetics
Visual Example of Genetics at Work
Dried and shrink-wrapped ear of corn illustrates naturally occurring phenotype ratios of genetic crosses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty