Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
3B Scientific® Numbered Skulls With Open Dentition
Supplier: AMERICAN 3B SCIENTIFIC SE
1020167 1020166
470002-702EA 353.4 USD InStock
470002-702 470118-896
3B Scientific® Numbered Skulls With Open Dentition
Models Human Biology Models
  • Includes 3B Smart Anatomy
This dental skull with opened mandible exposes tooth roots with vessels and nerves. The skull model also features the numbering of the cranial bones, bone components, fissures, foramina and other structures. The cranial sutures are shown in color, as are the meningeal vessels and venous sinuses. Not only are important anatomical features of the skull depicted but also the open jaw gives a distinctive view of teeth roots. The painted version includes muscle origins (red) and insertions (blue) represented on the left side of this human skull model.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty