Would you like to log in again?
3B Scientific® Lung Model With Larynx
Supplier: AMERICAN 3B SCIENTIFIC SE
1001243
470119-142EA 552.65 USD InStock
470119-142
3B Scientific® Lung Model With Larynx
Models Human Biology Models
  • 3 X Life Size
  • Base Represents The Diaphragm
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
This detailed heart depicts the structures of the diaphragm, the lungs, heart and larynx. The following parts of the heart can be removed: Esophagus, Trachea, Superior vena cava, Aorta, Pulmonary artery stem, Both atrium walls, Both ventricle walls.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty