Would you like to log in again?
3B Scientific® MRI Torso
Supplier: AMERICAN 3B SCIENTIFIC SE
1001237
470029-624EA 8450 USD InStock
470029-624
3B Scientific® MRI Torso
Models Human Biology Models
With this tool, a unique sectioning of the human body imitates MRIs in three dimensions.
  • Fifteen Sections
  • Three-Year Manufacturer's Warranty
  • Includes 3B Smart Anatomy
The model provides medical technology students with a better idea of how magnetic resonance tomography and ultrasound processes work. The torso is sliced horizontally from head to pelvis in 15 sections, providing a unique presentation that anatomy and pathology students will appreciate.
Anatomical structures are colored and detailed in relief on each part of the model. The sections, attached to a stand, can be rotated out one at a time or in groups for up-close comparative study.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty