Would you like to log in again?
45° and 60° Prism
Supplier: United Scientific Supplies
PATRAP
470106-852EA 15.8 USD InStock
470106-852
45° and 60° Prism
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate refracrion and internal reflection.
  • Clear acrylic
  • 15 mm thick
A clear acrylic trapezoid with a 45°, 60°, and parallel side featues one large flat surface, and is frosted to enhance visibility. The two bases measure 89 mm and 40 mm. It is 15 mm thick and 32 mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty