Would you like to log in again?
Adjustable Plant Light Stand
TT215WS
470014-972EA 146.95 USD InStock
470014-972
Adjustable Plant Light Stand
Lights Fluorescent Lights, Lamps and Bulbs
To start your seeds, and to keep them growing well, place them under this sturdy, compact plant light.
  • Easy To Assemble Or Store
  • Adjustable Light Height
  • Aluminum Base Resists Corrosion.
The housing holds two 15 W Gro-Lux plant light bulbs (included), and can be adjusted for a maximum height of 24" by simply moving the fixture up or down on the 1" square aluminum legs. Some assembly is required. Instructions are included.
Dimensions: 24″ H x 20″ W x 11.5″ D
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty