Would you like to log in again?
Allium, Leaf Epidermis Slide
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
WARD470177-830
470177-830EA 6.75 USD InStock
470177-830
Allium, Leaf Epidermis Slide
Slides Prepared Slides
Discover structures on a plant leaf surface.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows guard cells and stomata.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty