Would you like to log in again?
Aluminum Coated Pith-Like Balls
Supplier: SEMPCO INC. SE
BH-9107
470104-434PK 19.6 USD InStock
470104-434
Aluminum Coated Pith-Like Balls
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A thin aluminum coating produces great sensitivity to electrostatic charges. Styrofoam pith-like balls measure 8 mm in diameter and each has a silk suspension cord. Package of six.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty