Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Amoeba Model
Supplier: THE SCIENCE SOURCE SE
B136
470002-680EA 477.4 USD InStock
470002-680
Amoeba Model
Models Zoology Models
View The Same Structures As Living Organisms Without A Microscope
The 1,000X translucent model realistically portrays an amoeba in motion, with prominent pseudopods, representing natural movement. Sectioned to show internal cell details, the center of the amoeba clearly displays the nucleus, food vacuoles containing diatoms and bacteria, a fat droplet, contractile vacuole, and granular and clear zones of cytoplasm. The one-piece model is mounted on a base and includes a key, identifying 11 structures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty