Would you like to log in again?
Ward's® Amphibian Collection
Supplier: VWR
WARD470000-784
470000-784EA 58.49 USD InStock
470000-784
Ward's® Amphibian Collection
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved
With two types of salamanders, two types of frogs, and one toad, this set is ideal for studying the wide diversity of amphibians. Contents: Congo Eel; salamander or newt; bullfrog; grassfrog; and toad. The set of five is supplied in a single pail.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty