Would you like to log in again?
Animal Meiosis Model
Supplier: THE SCIENCE SOURCE SE
B139
470000-008EA 693.45 USD InStock
470000-008
Animal Meiosis Model
Models Molecular Biology Models
Each Stage Of Mitosis Has Its Own Model
The stages of meiosis are enlarged 1,600X and are highlighted in translucent “windows”, all on one colorful plaque. Each stage is connected by bold arrows so students can easily trace the steps in the process. The color scheme is similar to the Animal Mitosis model, so you can use both models to illustrate these two important processes of cell division. The plaque can be displayed in a prominent place in your classroom with the included stand. It comes with a key, identifying the stages of meiosis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty