Would you like to log in again?
Antacid Lab Activity
Supplier: Avantor
WARD470154-080
470154-080KT 123.99 USD InStock
470154-080
Antacid Lab Activity
Microchemistry Kits
Discover How Antacids Work
Using microchemistry techniques, students determine the neutralization and buffering characteristics of an over-the-counter antacid (included). Since all tests are performed using microscale techniques, only a small amount of test material is required. The kit contains three types of antacids, instructions, and materials for seven setups.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty