Would you like to log in again?
Aspirin Study Kit
Supplier: LAB-AIDS INC (KITS) SE
180
470150-746KT 152.75 USD InStock
470150-746
Aspirin Study Kit
Microchemistry Kits
This lab helps students become more informed consumers.
  • Activity: Testing and Comparing Different Aspirin Brands
  • Students Study Aspirin Brands for Filler, pH, and Per-Pill Cost
  • Process Informs Students of Ingredients in Common Products
  • Extension Activities Available
Aspirin (acetylsalicylic acid) might be the most used and abused non-prescription drug. Using aspirin tablets brought from home, students analyze and compare them with the Brand X aspirin included in this Lab-Aid.
They determine the percentage of aspirin and filler components in a tablet, check pH, and calculate the per-pill cost. Students draw conclusions on which aspirins are better, which are safer, and which do nothing at all. This consideration brings real-world questions into the science classroom. Extension activities are possible with this kit.
Ordering information: This lab includes Lugol's solution, Benedict's solution, iron nitrate solution, sodium carbonate solution, Brand X aspirin, dropper, sample bottles, test tubes, and blue litmus paper. Additional, various brands of aspirin are needed but not included.
Delivery information: Includes Teacher ‘s Guide and Student Copymasters
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty