Would you like to log in again?
Bacterial Morphology Composite Slides
Supplier: Avantor
WARD470177-400 WARD470180-850
470180-850EA 9.25 USD InStock
470180-850 470177-400
Bacterial Morphology Composite Slides
Slides Prepared Slides
Help your students learn about the different bacterial morphologies with side-by-side comparisons of pure cultures on the same slide.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Each slide contains a three separate smears of pure cultures of coccus, bacillus, and spirillum morphologies. Available in two different stain-types.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty