Would you like to log in again?
GREEN Basic Water Monitoring Kit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5886-20
470157-408CS 999.95 USD InStock
470157-408
GREEN Basic Water Monitoring Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
GREEN Basic Water Monitoring Kit features economical tests for eight different factors.
  • Material for 10 tests of eight factors
  • Developed by the Global Rivers Environmental Education Network
  • Test factors include coliform bacteria and nitrate
  • Kit includes bilingual manual
  • Aligned to NGSS MS-ESS3-4, MS-LS2-4, MS-LS2-5, HS-LS2-6, HS-ESS3-4
Using TesTabs, which are safe, nonhazardous reagent tablets, students can easily test for a variety of factors without risk. Developed by the Global Rivers Environmental Education Network, each monitoring kit comes with a bilingual (English and Spanish) manual.
This simple, safe, and economical package contains TesTabs for eight factors. Test factors included are biochemical oxygen demand, coliform bacteria, nitrate, dissolved oxygen, pH, phosphate, turbidity, and temperature change. It comes with material for ten tests each (three coliform tests).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty