Would you like to log in again?
Biology And Chemistry Of Soil Kit
Supplier: LAB-AIDS INC (KITS) SE
32
470004-612EA 158 USD InStock
470004-612
Biology And Chemistry Of Soil Kit
Standards Environmental Analysis Standards
Soil Feeds Us And Sustains Us.
  • Conduct A Thorough Evaluation Of Soil
  • Identify The Type And Variety Of Micro-Organisms (Soil Microbes)
  • Time Required: One 50 Minute Class Period
Time to play in the dirt! Students use ordinary soil samples to conduct a thorough evaluation of soil. Simple, easy-to-follow techniques and staining procedures are introduced early in this series of activities. After performing a series of activities, students are asked to draw conclusions concerning a soil’s ability to support plant growth. All tests are performed using either the patented Chemplate® or calibrated sample tubes.
Included:
Teacher’s Guide with SDS
Student Worksheets and Guides
Chemplates®, LAB-AIDS®
Cups, plastic
Plastic slides
Tubes, calibrated
Barium Chloride Solution, Drop control bottle
Phenolic Rose Bengal, Drop control bottle
pH Soil Indicator Solution, Drop control bottle
Potassium Dichromate Solution, Drop control bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty