Would you like to log in again?
Boreal Science Blood Typing Kit
Supplier: Avantor
WARD470104-036
470104-036KT 136.99 USD InStock
470104-036
Boreal Science Blood Typing Kit
Educational Classroom Kits and Activities
Students determine their own blood types with this easy to use kit. As this kit uses freeze-dried blood typing serum, it does not require refrigeration. Includes materials for 30 students, fliptop box, package of 50 sterile blood lancets, tallquist hemoglobin scale, freeze-dried blood typing serum, instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty