Would you like to log in again?
Braun Electroscope with Accessories
Supplier: VWR International
P45262
470149-478CS 1933 USD InStock
470149-478
Braun Electroscope with Accessories
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Versatile Apparatus for Electrostatics Lessons.
  • Includes instructions
This rugged, sensitive electroscope can accept various accessories to change its capacitance. A sensitive aluminum vane is balanced in a vertical position on a metal support which is insulated from its base. The vane is easily observed and projected on a chalkboard (Light Source, not included), on which an arbitrary scale has been marked. Height without attachments is about 9". Four attachments, each equipped with a banana plug which fits the vane system top, are included; a 3" conducting sphere; a proof plane with detachable insulating handle; and an aluminum cup (Faraday ice pail). Attachments can be charged using either our Electrophorus or our Power Supply (not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty