Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Breaking Board Paradox
Supplier: Avantor
WARD470152-848
470152-848EA 53.79 USD InStock
470152-848
Breaking Board Paradox
Educational Classroom Kits and Activities
This simple, effective demonstration lets students discover for themselves the paradoxes of Newton’s first and second laws.
A thin slab of white pine is set on a table so that a few inches hang over the edge. The board is then covered with a large sheet of paper that is subject to over 6000 kg of atmosphere because of its large surface area. When the overhanging portion of the board is hit sharply with the fist, it breaks easily. The other end of the board remains stationary, apparently held only by the sheet of paper. The kit contains instructions and materials for 10 demonstrations.
Caution: Note: Safety goggles should be worn and care should be taken to avoid flying pieces of wood when performing this experiment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty