Would you like to log in again?
Brine Shrimp Egg Hatchery
Supplier: SAN FRANCISCO BAY BRAND INC SE
66170-001
470149-872EA 26.85 USD InStock
470149-872
Brine Shrimp Egg Hatchery
Specimens Live Specimens Aquaria - Living Material
Keep A Living Food Source For Other Organisms In The Lab
  • Always have supply on hand
  • Comes with hatching chamber
Brine shrimp are an ideal food source for fish or Hydra; with this setup you can cultivate brine shrimp and always have a supply on hand. It comes with a black plastic hatching chamber with cover, plastic vial for transferring hatchlings from salt to fresh water, shrimp and salt mix, and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty