Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Buffers in Biology Lab Activity
Supplier: Avantor
WARD470012-254
470012-254EA 113.99 USD InStock
470012-254
Buffers in Biology Lab Activity
Educational Classroom Kits and Activities
Discover what buffers exist in potatoes and liver.
Students explore the buffering capabilities of plant and animal tissues you provide to determine the buffers that exist naturally in various substances and solutions. Through the investigation, students learn that living things have buffers in them in order to stabilize their pH. This understanding can then be related to the effect of fluctuating pH on enzyme activity and relate acid/base chemistry to anatomy and physiology. Developed by Julie Bohrer, the activity includes instructions. A blender and fresh biomaterials are required, but not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty