Would you like to log in again?
Bulk Spill Supplies
Supplier: CLIFFORD W. ESTES CO., INC.
06607
470145-380CS 75.25 USD InStock
470145-380
Bulk Spill Supplies
Spill Control Materials
Use inert, noncombustible vermiculite to contain solvent spills, sodium bicarbonate to neutralize acidic spills and waste, or sand to smother small fires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty