Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vernier® Bumper Launcher Kit
Supplier: VERNIER SOFTWARE & TECH SE
BLK
470017-530EA 98.99 USD InStock
470017-530
Vernier® Bumper Launcher Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Expand your physics experimentation capabilities.
  • Perform a wide range of experiments in dynamics and kinematics
This accessory kit to the Vernier Dual-Range Force Sensor and the Vernier Dynamics Cart and Track System is equipped with items that enable you to perform a wider range of experiments in dynamics and kinematics. Experiments can be conducted to investigate force during elastic collisions with or without contact, force during inelastic collisions, and work and energy to name a few.
Dynamics Track Bracket (1)
Dual-magnet bumper including a nylon bolt for securing magnetic bumper to DFS or the track bracket (1)
DFS end mount screw (1)
Single-magnetic bumpers (2)
Rubber bumpers (2)
Hoop bumpers (one stiff and one weak)(2)
Clay holders (2)
Clay (1)
NOTE: This kit contains very STRONG permanent magnets. Keep them away from objects that may be affected by strong magnetic fields.
For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty