Would you like to log in again?
C Clamp
Supplier: United Scientific Supplies
CLMPC3
470019-532EA 12.7 USD InStock
470019-532
C Clamp
Clamps General Purpose Clamps
Designed for clamping items together or securing parts to tabletops, the strong iron frame is also painted to reduce corrosion.
Nickel-plated screw and cross pin.
Jaw Opening: 3"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty