Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GPI Anatomicals® Canine Jaw Model
Supplier: GPI ANATOMICALS
9195
470029-460EA 166.95 USD InStock
470029-460
GPI Anatomicals® Canine Jaw Model
Models Zoology Models
Realistic Model Of Canine Dental Health
Open, close, and separate this average size canine jaw for up-close examination of normal dog teeth and common dental pathologies. While healthy teeth are depicted on the right side of the jaw, diseased and damaged teeth appear on the left, illustrating fractured canine, periodontal disease, tarter accumulation, plaque, gingivitis, worn incisors, retained deciduous tooth, gingival recession and missing premolar.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty