Would you like to log in again?
Celestial Spheres Kit
Supplier: SCIENCE INTERACTIVE GROUP
654-0030
470200-188PK 163 USD InStock
470200-188
Celestial Spheres Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
Present a Three-Dimensional Universe Model to Your Students
  • Model Daily Motion of Stars and Sun
  • Learn to use the sky for navigation
  • Teaches students the cause of seasons
Your students will be able to model the apparent daily motions of the stars and the Sun for a variety of latitudes or for any day of the year. With the included data, students will also be able to learn to use the sky for navigation and explain the cause of seasons on Earth. The kit can be used either in the laboratory or at home. It includes 20 clear plastic hemispheres, and ten of the following items: star charts with the 500 brightest stars in both the north and south hemispheres, marking pen, earth space globes, pins to represent the Sun, styrofoam bases, wooden dowels, plastic straws, instructions, and activities.
*Some experiments require students to be outside at sunset.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty