Would you like to log in again?
Ward's® Chemistry of Amino Acids and Proteins Lab Activity
Supplier: Avantor
WARD470007-972
470007-972KT 114.99 USD InStock
470007-972
Ward's® Chemistry of Amino Acids and Proteins Lab Activity
Educational Classroom Kits and Activities
Illustrate the Structure of a Protein in Three Dimensions
  • Kit can be reused each year
  • Students work together to form a polypeptide
The flexible plastic molecular components in this activity provide detail that no paper model can. Students learn about peptide bonds, protein structure, and protein synthesis as they explore first the general structure and composition of amino acids, and then link their amino acid models to see the structure of a polypeptide chain. Finally, the entire class can link their polypeptide chains together to form a protein molecule.
Ordering information: Kit includes Molecular Model Packets and Teacher Guide/Student Guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty