Would you like to log in again?
Ward's® Chemistry of Fats Lab Activity
Supplier: Avantor
WARD470007-970
470007-970KT 84.99 USD InStock
470007-970
Ward's® Chemistry of Fats Lab Activity
Educational Classroom Kits and Activities
Understand the Structure and Function of Fats
  • Understand the difference between saturated and unsaturated fats
  • Visually see how triglyceride synthesis occurs
As students construct models of glycerol and fatty acid sub-units and then put them together to form lipids, they will be able to visualize how triglyceride synthesis works. In the process, they will also learn the difference between saturated fats and unsaturated fats and how these two types of lipids can affect a healthy diet.
Includes:
Molecular Model Packets
Teacher Guide/Student Guide
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty