Would you like to log in again?
Ward's® Chick Development Museum Mount
Supplier: VWR
Ward's®
WARD470000-354
470000-354EA 1049.99 USD InStock
470000-354
Ward's® Chick Development Museum Mount
Specimens Preserved Specimens Display Specimens
Museum mounts are the ideal supplement for classroom dissection.
  • High quality display mount
  • Dissection reference for students
  • Quiz mechanism for teachers
Explain the development of this common domestic fowl in a few easy steps.
Museum Mounts are expertly crafted by our own Laboratory Technicians to show external and internal anatomy in great detail. Specimens are encased in acrylic with formalin for indefinite preservation.
Museum mounts can be used as a reference for teachers and students or as a quiz mechanism by covering up the key on the back of the case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty