Would you like to log in again?
Simulaids® Child Heart and Lungs Sounds Auscultation Trainer
Supplier: NASCO HEALTHCARE INC. SE
101-020
470125-150EA 3293.25 USD InStock
470125-150
Simulaids® Child Heart and Lungs Sounds Auscultation Trainer
Models Human Biology Models
Wirelessly Change The Sounds Students Hear
  • Speaker jack for full class listening
  • Can be used with any stethoscope
  • Includes case for storage or transport
This trainer emits the real-life recorded voice, lung, and heart sounds of a 4-year-old. The trainer operates from a wireless remote that has a simple turn-and-select method of choosing the sounds you wish to emulate and at what rate. The sounds are delivered to 10 lung and one heart speaker locations that can be auscultated with any stethoscope. This limits the special equipment that your students need and reduces costs. The trainer also has available a speaker jack through which you may broadcast the sounds being generated, so that the entire class can hear the same sounds. This makes it possible to utilize the trainer as a testing device for the entire class at once. No programming installation is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty