Would you like to log in again?
Ward's® Live Chlamydomonas reinhardtii mating set
Supplier: VWR
WARD470176-416
470176-416KT 29.95 USD InStock
470176-416
Ward's® Live Chlamydomonas reinhardtii mating set
Specimens Live Specimens Protists - Living Material
Demonstrates isogamous reproduction where gametes are similar in appearance and behavior. The included copymaster details asexual and sexual versions of the life cycle. Cultures include Chlamydomonas reinhardtii(+) and (-).
  • Strains Carefully Selected to Demonstrate isogamous reproduction
Make lectures and labs on protist reproduction clearer with our specially selected cultures that illustrate various reproductive phenomona. Set comes with mating strains of one type of protist, instructions, either the Working with Algae or the Working with Protozoa manual, and materials for 35 students.
Genus: Chlamydomonas Species: reinhardtii
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty