Would you like to log in again?
Ward's® Chromatography of Algae Pigments Lab Activity
Supplier: Avantor
470030-206KT 127.79 USD InStock
470030-206
Ward's® Chromatography of Algae Pigments Lab Activity
Educational Classroom Kits and Activities
Examine Algae Pigments Using Thin Layer Chromatography
By separating the pigments from three different algae types that are supplied, students can then analyze the results to study the differences in the pigments used to classify algae types. The set includes: red algae, brown algae, and green algae cultures; a teacher’s manual; student copymaster; and enough materials for six setups. Note: This activity must be performed in a well ventilated area or under a fume hood. Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty