Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Chromatography of Spinach Kit
Supplier: Avantor
WARD470024-176
470024-176KT 75.29 USD InStock
470024-176
Ward's® Chromatography of Spinach Kit
Educational Classroom Kits and Activities
This kit produces colorful results in just fifteen minutes.
  • Students research different affinities for mobile and stationary phases
  • Pre- and post-laboratory assessments included
  • Fast-acting experiment
  • Material for 15 setups
  • Estimated lab time: 45 minutes
The procedure in this activity quickly yields large amounts of plant pigments, making it easier to complete than traditional solvent extractions. Students learn the basics of paper chromatography, including the use of petroleum ether or acetone solvent, and calculate the rate of flow (Rƒ) for each pigment.
Once students complete the experiment, they are rewarded with a vivid chromatogram that displays chlorophyll A and B, xanthophyll, and carotene pigments. They also gain an understanding of the different affinities for mobile and stationary phases. The kit includes a teacher’s guide, a student copymaster, and enough material for 15 setups. It is recommended that this procedure be performed under a fume hood.
Ordering information: This kit includes chromatography vials with caps, chromatography strips, disposable pipets, metric rulers, and chromatography solvent. Fresh spinach leaves, coins, pencils, scissor, forceps, and a chemical fume hood are required but not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty