Would you like to log in again?
Classroom Mineral Collection
Supplier: VWR
WARD470016-498
470016-498KT 147.59 USD InStock
470016-498
Classroom Mineral Collection
Specimens Mineral Specimens Mineral Collections
The Classroom Mineral Collection is a fascinating and colorful classroom study set.
  • Compartmented Collection Box
  • Great for All Levels of Teaching
  • Identification Guide Included
  • Thirty-Six Numbered Samples
The included minerals, collected from around the world, are ideal for introducing students to Mohs' hardness testing, basic rock-forming minerals, and ores used in important products.
The set is housed in a compartmented collection box.
Ordering information: This kit includes 36 numbered samples (1×2"): 9 hardness test minerals, 15 common rock-forming minerals, and 12 economic minerals and ores.
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty