Would you like to log in again?
Classroom Mineral Collection
Supplier: VWR
WARD470015-804
470015-804EA 115.79 USD InStock
470015-804
Classroom Mineral Collection
Specimens Mineral Specimens Mineral Collections
A fascinating and colorful classroom study set, the included minerals, collected from around the world, are ideal for introducing students to Mohs' hardness testing, basic rock-forming minerals and minerals and ores used in products important to our economy. The set is housed in a compartmented collection box.
Delivery information: Includes 36 numbered samples (1×2"): 9 Hardness Test Minerals, 15 Common Rock-Forming Minerals and 12 Economic Minerals and Ores.
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty