Would you like to log in again?
Ward's® Geo-Logic Cleavage, Fracture, and Parting Collection
Supplier: VWR
WARD470025-338
470025-338EA 65.09 USD InStock
470025-338
Ward's® Geo-Logic Cleavage, Fracture, and Parting Collection
Specimens Mineral Specimens Mineral Collections
Examples of Different Forms of Breakage in Minerals
  • Developed by teachers and WARD's Staff Geologists
  • Classroom tested for dependabilty
  • Designed to save valuble planning and preparation time
Demonstrate the structure of minerals with specimens known for well-defined breaking patterns in our GEO-logic collection. A teacher's guide and student copymasters are included for the following activities: internal structure of minerals — how they break; cleavage directions and measurement of angles; and significance of cleavage, fracture, and parting. The set is housed in a compartmented collection box.
The GEO-logic system provides a modular approach to teaching and allows you to address broad curriculum needs for the study of minerals and rocks; natural resources and energy; and fossils and earth history. The types of GEO-logic teaching collections include:
• Topic Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters
• Classroom Bundles, which provide 15 bulk-packaged samples included in the reference or topic set and 15 sets of any required testing materials included with the corresponding set
*Classroom Bundles are bulk-packaged and do not include display boxes, or teacher/student activity guides.
Contents:12 numbered samples, 1"" x 2""; magnifier; protractor; ruler
Minerals: biotite, calcite, corundum, fluorite, garnet, halite, hornblende, magnetite, microcline, quartz, sphalerite, tremolite
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty