Would you like to log in again?
Coleslaw Chemistry Kit
Supplier: Avantor
WARD470152-860
470152-860KT 98.69 USD InStock
470152-860
Coleslaw Chemistry Kit
Educational Classroom Kits and Activities
Use Cabbage Juice as a pH Indicator
  • Ideal for Grades 8-11
  • Create Your Own pH Reference Chart
  • 5 Activities Included
This unique chemistry lab is based on the properties of red cabbage juice as a pH indicator. Students make indicator solution and pH strips out of red cabbage juice and then use the strips to create their own pH reference chart. Students test the acidity of "rain" in a controlled atmosphere.
Includes:
Forceps
Safety Matches
3" x 5" Index Cards
Bags
Filter Paper Strips
Glass Plates
Graduated Pipettes
Spot Plates
Buffer Capsules - pH 3 - 11
Instructions
Required but Not Provided: Head of Red Cabbage, Distilled Water, Container to boil cabbage, Stove or hot plate, Knife, Cutting Board, Scissors, Beakers, Dropping Bottles (optional), Colored Pencils or Crayons, Permanent Markers, Plastic Strainer, Vials or ice cube trays, Erlenmeyer flasks with stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty