Would you like to log in again?
Color Filter Set
Supplier: Avantor
WARD470002-766
470002-766KT 15.4 USD InStock
470002-766
Color Filter Set
Educational Materials Physics Educational Materials Optics Learning Activities
Explore The Absorption of Light
  • Comes in a convenient storage box!
Each acrylic filter measures 2" square and 1/8" thick. The set includes one each of blue, green, yellow, orange, and red filters
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty