Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Colossus Floor Case

Supplier: WADDELL CO THE SE
2605-PB-SN
470002-322EA 6090.4 USD InStock
470002-322
Colossus Floor Case
Furniture Casework
This durable case securely exhibits science displays.

  • Width: 60"
  • Height: 66"
  • Diameter: 20"
  • Weight: 362 lb.
  • Built-In Levers

This metal case has built-in levers and eight half-length glass shelves.

It is available with a bronze, satin aluminum, or gold finish and white laminate or plaque fabric back.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty