Would you like to log in again?
Common West Coast Sea Urchin
Supplier: VWR
WARD470000-774
470000-774PK 26.95 USD InStock
470000-774
Common West Coast Sea Urchin
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved
Strongylocentrotus purpuratus. Body size: 1–2".
Characterized by complex feeding structures with five calcareous protractable teeth (Aristotle’s lantern).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty