Would you like to log in again?
Ward's® Complete Petro-Lab Study Set
Supplier: VWR
470233-426EA 1366.3 USD InStock
470233-426
Ward's® Complete Petro-Lab Study Set
Specimens Rock Specimens Rock Collections
Combine all six sets and save!
Special series designed for introductory college Petrography labs. Each thin section set contains seven slides illustrating important mineral associations, textural features, and optical properties. Major mineralogical features are described in a worksheet that accompanies each slide. Using the basic information provided, students can determine optical properties, percentage estimations, and general compositional features that will assist them in their recognition of rocks and minerals.
This collection contains all 21 thin section slides, as well as all 21 matching hand specimens.
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty