Would you like to log in again?
Composition of Soil Kit
Supplier: THE SCIENCE SOURCE SE
2000
470112-650KT 290 USD InStock
470112-650
Composition of Soil Kit
Educational Materials Environmental Educational Materials Environmental Learning Activities
What chemicals make up your local soil and why are they so important to living things found in the soil? Students will answer these questions and many more with this compact, teacher-designed soil test kit.
  • Students can work cooperatively
  • Discussion questions assess understanding
  • Tests several soil quality factors
Simple chemical testing procedures are employed to determine water percentage; soil organic content; pH; reagent tests for calcium, carbonates, magnesium, phosphates and sulfates; and test paper determinations for ammonium, iron, nitrate and potassium. Discussion questions are furnished for each component. Includes enough materials for 30 students working in pairs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty