Would you like to log in again?
Ward's® Constructing a Dichotomous Key Lab Activity
Supplier: Avantor
WARD470015-354
470015-354KT 76.49 USD InStock
470015-354
Ward's® Constructing a Dichotomous Key Lab Activity
Educational Classroom Kits and Activities
Learn About Dichotomous Keys by Classifying Seeds
Introduce your students to the most commonly used binary system of classification — the dichotomous key. Students will create their own key by using observation and clear descriptors in order to accurately categorize and identify seeds. Once they have finished their own key, students will exchange keys for a peer review to ensure that each organism can be properly identified. The kit includes materials for 15 setups, a teacher’s guide, and student copymaster.
Materials Included
•1 Package, sweet corn seeds
•1 Package, wrinkled pea seeds
•2 Packages, sunflower seeds
•1 Package, wheat seeds
•1 Package, oat seeds
•1 Package, black-eyed peas
•1 Package, mung beans
•1 Package, kidney beans
•200 Small zipper bags
•15 Large zipper bags
•15 Metric rulers
Materials Needed But Not Provided
•Scissors
•Paper
•Pen or pencil
•Permanent marker
Time Requirement
•Part A: 15 Minutes
•Part B: 45 Minutes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty